Gene Transcripts

Gene Symbol
Mroh2b
Entrez Gene ID
361902
Gene Name
maestro heat-like repeat family member 2B
Gene Aliases
Heatr7b2, RGD1305413
Chromosome Location
Chr. 2: 54621176 - 54686155 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760805.1 XP_008759027.1 30 3229

View Full Product Details