Gene Transcripts

Gene Symbol
Tfap2c
Entrez Gene ID
362280
Gene Name
transcription factor AP-2 gamma
Gene Aliases
AP2-gamma, Tcfap2c
Chromosome Location
Chr. 3: 170550314 - 170559202 on Build Rnor_6.0
UniGene ID
Rn.16142
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201420.2 NP_958823.2 3 764
XM_006235691.2 XP_006235753.1 3 733
GenBank mRNA BC097260.1 - 3 764

View Full Product Details