Gene Transcripts

Gene Symbol
Anln
Entrez Gene ID
363031
Gene Name
anillin, actin binding protein
Gene Aliases
RGD1561367
Chromosome Location
Chr. 8: 23336455 - 23397115 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766019.1 XP_008764241.1 2 343
XM_008766020.1 XP_008764242.1 2 343

View Full Product Details