Gene Transcripts

Gene Symbol
Sp9
Entrez Gene ID
366078
Gene Name
trans-acting transcription factor 9
Gene Aliases
RGD1559457
Chromosome Location
Chr. 3: 59981959 - 59984489 on Build Rnor_6.0
UniGene ID
Rn.228204
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191902.1 NP_001178831.1 2 786

View Full Product Details