Gene Transcripts

Gene Symbol
Scrt2
Entrez Gene ID
366229
Gene Name
scratch family zinc finger 2
Gene Aliases
RGD1564796
Chromosome Location
Chr. 3: 147585947 - 147597660 on Build Rnor_6.0
UniGene ID
Rn.228224
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191905.1 NP_001178834.1 1, 2 137

View Full Product Details