Gene Transcripts

Gene Symbol
Six2
Entrez Gene ID
366542
Gene Name
SIX homeobox 2
Gene Aliases
Chromosome Location
Chr. 6: 8952572 - 8955893 on Build Rnor_6.0
UniGene ID
Rn.225521
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191908.1 NP_001178837.1 2 604

View Full Product Details