Gene Transcripts

Gene Symbol
G4
Entrez Gene ID
406868
Gene Name
G4 protein
Gene Aliases
Chromosome Location
Chr. 20: 5117991 - 5120332 on Build Rnor_6.0
UniGene ID
Rn.205352
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003975.3 NP_001003975.1 3 1279
XM_006256073.2 XP_006256135.1 2 1490
GenBank mRNA BC101859.1 - 3 1259

View Full Product Details