Gene Transcripts

Gene Symbol
Ka11
Entrez Gene ID
450226
Gene Name
type I keratin KA11
Gene Aliases
Chromosome Location
Chr. 10: 88141667 - 88144624 on Build Rnor_6.0
UniGene ID
Rn.145402
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008750.1 NP_001008750.1 2 533

View Full Product Details