Gene Transcripts

Gene Symbol
Vom1r74
Entrez Gene ID
494262
Gene Name
vomeronasal 1 receptor 74
Gene Aliases
V1rc16
Chromosome Location
Chr. 4: 87814741 - 87815652 on Build Rnor_6.0
UniGene ID
Rn.129666
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008911.1 NP_001008911.1 1 692
GenBank mRNA AY510285.1 - 1 692

Gene Symbol
LOC100909399
Entrez Gene ID
100909399
Gene Name
vomeronasal type-1 receptor A16-like
Gene Aliases
Chromosome Location
Chr. Un|NW_007906889: 126808 - 127719 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749767.1 XP_003749815.1 1 692

View Full Product Details