Gene Transcripts

Gene Symbol
Vom1r50
Entrez Gene ID
494267
Gene Name
vomeronasal 1 receptor 50
Gene Aliases
V1rg11
Chromosome Location
Chr. 9: 52400683 - 52401660 on Build Rnor_6.0
UniGene ID
Rn.129417
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008918.1 NP_001008918.1 441
GenBank mRNA AY510350.1 - 441

View Full Product Details