Gene Transcripts

Gene Symbol
Vom1r24
Entrez Gene ID
494283
Gene Name
vomeronasal 1 receptor 24
Gene Aliases
V1re14
Chromosome Location
Chr. 8: 3638921 - 3639871 on Build Rnor_6.0
UniGene ID
Rn.129442
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008936.1 NP_001008936.1 269
GenBank mRNA AY510362.1 - 269

View Full Product Details