Gene Transcripts

Gene Symbol
Vom1r31
Entrez Gene ID
494312
Gene Name
vomeronasal 1 receptor 31
Gene Aliases
V1re12
Chromosome Location
Chr. Un|NW_007906935: 24525 - 25457 on Build Rnor_6.0
UniGene ID
Rn.129433
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008969.1 NP_001008969.1 222
GenBank mRNA AY510343.1 - 222

View Full Product Details