Gene Transcripts

Gene Symbol
Sco1
Entrez Gene ID
497930
Gene Name
SCO1 cytochrome c oxidase assembly protein
Gene Aliases
RGD1559538
Chromosome Location
Chr. 10: 53595854 - 53608437 on Build Rnor_6.0
UniGene ID
Rn.203819
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173374.1 NP_001166845.1 6 799
GenBank mRNA BC158882.1 - 6 734

View Full Product Details