Gene Transcripts

Gene Symbol
Gal3st4
Entrez Gene ID
498166
Gene Name
galactose-3-O-sulfotransferase 4
Gene Aliases
RGD1561105
Chromosome Location
Chr. 12: 19583110 - 19592440 on Build Rnor_6.0
UniGene ID
Rn.48145
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109064.1 NP_001102534.1 4 1477
XM_006249025.2 XP_006249087.1 4 1709
XM_006249026.2 XP_006249088.1 4 1495

View Full Product Details