Gene Transcripts

Gene Symbol
Otud1
Entrez Gene ID
498803
Gene Name
OTU deubiquitinase 1
Gene Aliases
RGD1563344
Chromosome Location
Chr. 17: 86406647 - 86410670 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_574086.6 XP_574086.3 1 2719

View Full Product Details