Gene Transcripts

Gene Symbol
Mtcp1
Entrez Gene ID
498814
Gene Name
mature T-cell proliferation 1
Gene Aliases
RGD1561352
Chromosome Location
Chr. 18: 411159 - 411923 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_574098.4 XP_574098.2 2 275

View Full Product Details