Gene Transcripts

Gene Symbol
Brsk1
Entrez Gene ID
499073
Gene Name
BR serine/threonine kinase 1
Gene Aliases
RGD1563268, Sad-b
Chromosome Location
Chr. 1: 72701281 - 72727061 on Build Rnor_6.0
UniGene ID
Rn.112844
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127337.1 NP_001120809.1 14 1647
XM_006228283.1 XP_006228345.1 13 1512
GenBank mRNA AB365521.1 - 14 1647

View Full Product Details