Gene Transcripts

Gene Symbol
Dkkl1
Entrez Gene ID
499150
Gene Name
dickkopf-like 1
Gene Aliases
RGD1563523
Chromosome Location
Chr. 1: 101205426 - 101210675 on Build Rnor_6.0
UniGene ID
Rn.125407
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109145.1 NP_001102615.1 3 647

View Full Product Details