Gene Transcripts

Gene Symbol
Fxn
Entrez Gene ID
499335
Gene Name
frataxin
Gene Aliases
RGD1565754
Chromosome Location
Chr. 1: 242123975 - 242153168 on Build Rnor_6.0
UniGene ID
Rn.13133
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191952.1 NP_001178881.1 2 180
XM_008760308.1 XP_008758530.1 2 514

View Full Product Details