Gene Transcripts

Gene Symbol
Plekhd1
Entrez Gene ID
500685
Gene Name
pleckstrin homology domain containing, family D (with coiled-coil domains) member 1
Gene Aliases
RGD1566112
Chromosome Location
Chr. 6: 104340114 - 104371162 on Build Rnor_6.0
UniGene ID
Rn.21179
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127566.1 NP_001121038.1 2 373
GenBank mRNA BC161896.1 - 2 373

View Full Product Details