Gene Transcripts

Gene Symbol
Ttc9
Entrez Gene ID
500689
Gene Name
tetratricopeptide repeat domain 9
Gene Aliases
RGD1565985
Chromosome Location
Chr. 6: 105364668 - 105399500 on Build Rnor_6.0
UniGene ID
Rn.8149
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134731.1 NP_001128203.1 2 498
GenBank mRNA BC166802.1 - 2 424

View Full Product Details