Gene Transcripts

Gene Symbol
Mettl5
Entrez Gene ID
502632
Gene Name
methyltransferase like 5
Gene Aliases
RGD1566062
Chromosome Location
Chr. 3: 56101597 - 56113823 on Build Rnor_6.0
UniGene ID
Rn.117034
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114181.1 NP_001107653.1 5, 6 755
GenBank mRNA BC158754.1 - 5, 6 755

View Full Product Details