Gene Transcripts

Gene Symbol
Defb10
Entrez Gene ID
641635
Gene Name
defensin beta 10
Gene Aliases
Chromosome Location
Chr. 16: 75198178 - 75208230 on Build Rnor_6.0
UniGene ID
Rn.159994
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037510.1 NP_001032599.1 2 105
GenBank mRNA AY621342.1 - 2 105

View Full Product Details