Gene Transcripts

Gene Symbol
Mbd4
Entrez Gene ID
680915
Gene Name
methyl-CpG binding domain protein 4
Gene Aliases
Chromosome Location
Chr. 4: 147744673 - 147756452 on Build Rnor_6.0
UniGene ID
Rn.179073
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001059437.5 XP_001059437.3 2 414
GenBank mRNA BC158892.1 - 2 235

View Full Product Details