Gene Transcripts

Gene Symbol
Prl5a2
Entrez Gene ID
691469
Gene Name
prolactin family 5, subfamily a, member 2
Gene Aliases
Prlpp
Chromosome Location
Chr. 17: 38456016 - 38462358 on Build Rnor_6.0
UniGene ID
Rn.195313
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044702.1 NP_001038167.1 2 235
GenBank mRNA DQ329280.1 - 2 235

View Full Product Details