Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD40
Entrez Gene ID
706451
Gene Name
ankyrin repeat domain 40
Gene Aliases
-
Chromosome Location
Chr.16: 46079057 - 46093698 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015119268.2 XP_014974754.2 - 1020 87
GenBank mRNA JU337053.1 - - 828 87
JU477781.1 - - 960 87
JV048657.1 - - 966 87
JV598031.1 - - 1018 87
JV598032.1 - - 1018 87

View Full Product Details