The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_015112712.2 | XP_014968198.1 | - | 807 | 66 |
XM_028832358.1 | XP_028688191.1 | - | 807 | 66 | |
GenBank mRNA | JV642400.1 | - | - | 38 | 66 |