Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNPY4
Entrez Gene ID
719080
Gene Name
canopy FGF signaling regulator 4
Gene Aliases
EGK_13514
Chromosome Location
Chr.3: 42302272 - 42307826 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266755.1 NP_001253684.1 - 670 90
XM_028845070.1 XP_028700903.1 - 595 90
GenBank mRNA JU333765.1 - - 682 90
JU475011.1 - - 628 90
JV046634.1 - - 670 90

View Full Product Details