Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R21
Entrez Gene ID
715917
Gene Name
protein phosphatase 1 regulatory subunit 21
Gene Aliases
EGK_05315, KLRAQ1
Chromosome Location
Chr.13: 60230960 - 60323275 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015112318.2 XP_014967804.1 - 1620 99
XM_015112321.2 XP_014967807.1 - 1656 99
XM_028831687.1 XP_028687520.1 - 1469 99
XM_028831688.1 XP_028687521.1 - 1603 99
XM_028831689.1 XP_028687522.1 - 1465 99
XM_028831690.1 XP_028687523.1 - 1642 99
XM_028831691.1 XP_028687524.1 - 1409 99
GenBank mRNA JU332921.1 - - 1552 99
JU474323.1 - - 1510 99
JV706950.1 - - 1574 99

View Full Product Details