Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PFDN6
Entrez Gene ID
718253
Gene Name
prefoldin subunit 6
Gene Aliases
EGK_14781
Chromosome Location
Chr.4: 136215628 - 136217497 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194865.1 NP_001181794.1 - 181 92
XM_015136071.2 XP_014991557.1 - 334 92
XM_028846862.1 XP_028702695.1 - 375 92
GenBank mRNA JU332517.1 - - 359 92
JU473993.1 - - 364 92
JV045918.1 - - 196 92

View Full Product Details