Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL32
Entrez Gene ID
709276
Gene Name
kelch like family member 32
Gene Aliases
-
Chromosome Location
Chr.4: 76157820 - 76376594 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261738.1 NP_001248667.1 - 2025 113
XM_015136955.2 XP_014992441.1 - 7411 113
XM_028846771.1 XP_028702604.1 - 2138 113
XM_028846772.1 XP_028702605.1 - 2083 113
XM_028846773.1 XP_028702606.1 - 3120 113
XM_028846774.1 XP_028702607.1 - 2292 113
GenBank mRNA JU321841.1 - - 2025 113

View Full Product Details