Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CMBL
Entrez Gene ID
696762
Gene Name
carboxymethylenebutenolidase homolog
Gene Aliases
EGK_16358
Chromosome Location
Chr.6: 10249372 - 10279882 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193768.2 NP_001180697.1 - 500 84
XM_015139728.2 XP_014995214.1 - 307 84
GenBank mRNA JU323333.1 - - 466 84
JU473785.1 - - 454 84
JV045080.1 - - 442 84

View Full Product Details