Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FUT11
Entrez Gene ID
706552
Gene Name
fucosyltransferase 11
Gene Aliases
EGK_19725
Chromosome Location
Chr.9: 64027249 - 64035625 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001100876.3 XP_001100876.1 - 877 61
XM_002805680.3 XP_002805726.1 - 877 61
XM_015147295.2 XP_015002781.1 - 877 61
GenBank mRNA JU325993.1 - - 150 61
JV644112.1 - - 708 61

View Full Product Details