Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KERA
Entrez Gene ID
709743
Gene Name
keratocan
Gene Aliases
-
Chromosome Location
Chr.11: 90823851 - 90831703 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001102581.4 XP_001102581.2 - 611 64

View Full Product Details