Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCFC2
Entrez Gene ID
716061
Gene Name
GC-rich sequence DNA-binding factor 2
Gene Aliases
C2orf3
Chromosome Location
Chr.13: 33014971 - 33065903 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194347.3 NP_001181276.3 - 1954 96
XM_015112563.2 XP_014968049.2 - 2531 96
XM_028831625.1 XP_028687458.1 - 2462 96
GenBank mRNA JU325969.1 - - 872 96

View Full Product Details