Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEX37
Entrez Gene ID
699461
Gene Name
testis expressed 37
Gene Aliases
C13H2orf51, C2orf51, EGK_05546
Chromosome Location
Chr.13: 18451302 - 18456306 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193977.3 NP_001180906.1 - 181 69
XM_015112802.2 XP_014968288.1 - 200 69

View Full Product Details