Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFAF5
Entrez Gene ID
694292
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 5
Gene Aliases
C10H20orf7, EGK_02679
Chromosome Location
Chr.10: 52728716 - 52764370 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001083688.4 XP_001083688.1 - 978 133
XM_028828171.1 XP_028684004.1 - 861 133
XM_028828173.1 XP_028684006.1 - 848 133
XM_028828174.1 XP_028684007.1 - 663 133
GenBank mRNA JU323224.1 - - 887 133
JU472008.1 - - 887 133
JV635786.1 - - 898 133
JV704659.1 - - 887 133

View Full Product Details