Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIMT1
Entrez Gene ID
696688
Gene Name
DIMT1 rRNA methyltransferase and ribosome maturation factor
Gene Aliases
DIMT1L, EGK_16525, HSA9761
Chromosome Location
Chr.6: 60786357 - 60800488 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015140094.2 XP_014995580.1 - 846 121
XM_028849454.1 XP_028705287.1 - 928 121

View Full Product Details