Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BEND6
Entrez Gene ID
712498
Gene Name
BEN domain containing 6
Gene Aliases
EGK_15041
Chromosome Location
Chr.4: 112545400 - 112620334 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257859.2 NP_001244788.1 - 684 73
XM_015136544.2 XP_014992030.1 - 430 73
XM_015136545.2 XP_014992031.1 - 423 73
XM_028846798.1 XP_028702631.1 - 1590 73
XM_028846799.1 XP_028702632.1 - 424 73
GenBank mRNA JU333401.1 - - 684 73
JV608936.1 - - 699 73

View Full Product Details