Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT10B
Entrez Gene ID
708906
Gene Name
Wnt family member 10B
Gene Aliases
-
Chromosome Location
Chr.11: 48573126 - 48587984 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194299.2 NP_001181228.2 - 528 62
XM_015151618.2 XP_015007104.2 - 1749 62
XM_015151620.2 XP_015007106.2 - 1455 62
XM_015151621.2 XP_015007107.2 - 1751 62
GenBank mRNA JU321571.1 - - 466 62
JV565979.1 - - 484 62
JV600669.1 - - 518 62

View Full Product Details