Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESX1
Entrez Gene ID
699644
Gene Name
ESX homeobox 1
Gene Aliases
EGK_20785
Chromosome Location
Chr.X: 100911797 - 100916776 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001092145.4 XP_001092145.1 - 642 63
GenBank mRNA JV662311.1 - - 128 63

View Full Product Details