Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A31
Entrez Gene ID
710042
Gene Name
solute carrier family 25 member 31
Gene Aliases
EGK_16082
Chromosome Location
Chr.5: 125503908 - 125536702 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194300.2 NP_001181229.1 - 895 86
GenBank mRNA JV072776.1 - - 633 86
JV635675.1 - - 898 86

View Full Product Details