Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NELFCD
Entrez Gene ID
696053
Gene Name
negative elongation factor complex member C/D
Gene Aliases
TH1L
Chromosome Location
Chr.10: 94143747 - 94157496 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015148701.2 XP_015004187.1 - 156 99
XM_028828541.1 XP_028684374.1 - 156 99
GenBank mRNA JU487950.1 - - 86 99
JV666253.1 - - 109 99

View Full Product Details