Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARSI
Entrez Gene ID
712077
Gene Name
arylsulfatase family member I
Gene Aliases
EGK_17023
Chromosome Location
Chr.6: 147809722 - 147816332 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001108178.4 XP_001108178.1 - 888 151

View Full Product Details