Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYGB
Entrez Gene ID
707130
Gene Name
cytoglobin
Gene Aliases
-
Chromosome Location
Chr.16: 72931513 - 72941153 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257930.1 NP_001244859.1 - 631 79
GenBank mRNA JU332798.1 - - 631 79
JV046109.1 - - 585 79
JV639122.1 - - 625 79

View Full Product Details