Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLX1
Entrez Gene ID
695620
Gene Name
distal-less homeobox 1
Gene Aliases
-
Chromosome Location
Chr.12: 59202959 - 59207128 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260619.1 NP_001247548.1 - 510 95
GenBank mRNA JU333234.1 - - 510 95

View Full Product Details