Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO40
Entrez Gene ID
713933
Gene Name
F-box protein 40
Gene Aliases
-
Chromosome Location
Chr.2: 154706392 - 154743318 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001111331.4 XP_001111331.2 - 2269 72
XM_015129973.2 XP_014985459.2 - 2371 72
XM_015129974.2 XP_014985460.2 - 1971 72
XM_015129976.2 XP_014985462.2 - 2234 72
XM_015129977.2 XP_014985463.2 - 2054 72

View Full Product Details