Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DESI2
Entrez Gene ID
713515
Gene Name
desumoylating isopeptidase 2
Gene Aliases
PPPDE1
Chromosome Location
Chr.1: 4401240 - 4464048 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257794.1 NP_001244723.1 - 225 116
GenBank mRNA JU334765.1 - - 225 116
JU475806.1 - - 264 116
JV652911.1 - - 271 116

View Full Product Details