The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257607.1 | NP_001244536.1 | - | 740 | 74 |
XM_015110368.2 | XP_014965854.2 | - | 1467 | 74 | |
XM_015110369.2 | XP_014965855.2 | - | 1120 | 74 | |
XM_028830543.1 | XP_028686376.1 | - | 570 | 74 | |
GenBank mRNA | JU334668.1 | - | - | 740 | 74 |
JU475716.1 | - | - | 740 | 74 | |
JV047161.1 | - | - | 739 | 74 |