Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ETNK2
Entrez Gene ID
700683
Gene Name
ethanolamine kinase 2
Gene Aliases
-
Chromosome Location
Chr.1: 61638546 - 61659360 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260518.1 NP_001247447.1 - 765 62
XM_015118445.2 XP_014973931.1 - 969 62
XM_015118477.2 XP_014973963.1 - 709 62
XM_028835182.1 XP_028691015.1 - 969 62
XM_028835302.1 XP_028691135.1 - 969 62
XM_028835329.1 XP_028691162.1 - 969 62
GenBank mRNA JU325863.1 - - 456 62
JV047529.1 - - 765 62
JV655197.1 - - 936 62

View Full Product Details